Multiple affinity removal starter reagent kits
Multiple affinity removal buffer A, 1 L, For loading, washing, equilibrating, 1
Codice catalogo AGLS5185-5987
Nome del fornitore: AGILENT
Multiple affinity removal starter reagent kits
Codice catalogo AGLS5185-5987
Nome del fornitore: AGILENT
Numero CAS:
Vai a
Product Details & Documents
Starter reagent kit for multiple affinity removal LC columns.
Informazioni per l'ordine: Includes: 2 bottles buffer A (1 L), 1 bottle buffer B (1 L), 2 packs of 25 spin filters, 1 pack of 25 spin concentrator.
Specifiche
Specifiche
- Conf:1
- Descrizione:Multiple affinity removal buffer A, 1 L
- Details:For loading, washing, equilibrating