Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Friedman-Pearson rongeurs

Friedman-Pearson rongeurs, 0,5 mm cup diameter, Straight, 140 mm
Codice catalogo FSCI16220-14
Marketsource
Nome del fornitore:   FINE SCIENCE TOOLS
Friedman-Pearson rongeurs
Codice catalogo FSCI16220-14
Nome del fornitore:  FINE SCIENCE TOOLS
Marketsource
Friedman-Pearson rongeurs
Friedman-Pearson rongeurs

Friedman-Pearson rongeurs, 0,5 mm cup diameter, Straight, 140 mm

Price
SFr. 287.66
Log in per disponibilità
1 * 1 ST
Product Details & Documents

These rongeurs are made of stainless steel.

  • Single joint action
Specifiche
  • Tipo:
    Straight
  • Descrizione:
    Friedman-Pearson rongeurs, 0,5 mm cup diameter
  • Lunghezza:
    140 mm
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.