Polypropylene Muff Covers with Elastic
Polypropylene muff cover, 4" (102 mm), fits 1 - 1,75" (25,5 - 44,5 mm) Ø, bianco
Codice catalogo 129-3067
Nome del fornitore: KEYSTONE CLEANROOM PRODUCTS
Polypropylene Muff Covers with Elastic
Codice catalogo 129-3067
Nome del fornitore: KEYSTONE CLEANROOM PRODUCTS
Vai a
Product Details & Documents
Keeps cleaned non sterile production hoses protected from external sources of contamination.
- Manufactured and packed in a controlled cleanroom environment
- Reduces labour costs
- Tear and puncture resistant
- Conveniently packaged in 100 per sealed poly bag
These polypropylene muffs are a great choice in non sterile pharmaceutical manufacturing operations. Can be used as a clean equipment cover to protect process hoses.
Poly bagged in packs of 100 to help ensure cleanliness up until point of use.
Informazioni per l'ordine: Certificates of Conformance sent with each shipment.
Specifiche
- Colore:bianco
- Descrizione:Polypropylene muff cover, 4" (102 mm), fits 1 - 1,75" (25,5 - 44,5 mm) Ø
- Seq:1