Bucket Liners
BucketLiner™, 3 mil, fits B-99 buckets, LDPE/nylon, 889×533 mm (35×21") mm
Codice catalogo MICRBKL99-3521
Nome del fornitore: MICRONOVA
Vai a
Product Details & Documents
The unique LDPE/nylon laminate is a good barrier material for transporting raw materials or to line buckets containing disinfectants or other cleaning solutions.
- Laminate is 75% LDPE, 25% nylon
- Excellent for disinfectants and cleaning solutions
- Ideal for product parts transport
Designed for cleanrooms and controlled environments. Available as sterile and non-irradiated.
Packaging: Individually double packed, packed in a case of 24/50/200.
Documents
Specifiche
- Descrizione:BucketLiner™, 3 mil, fits B-99 buckets
- L×L:889×533 mm (35×21") mm
- Materiale:LDPE/nylon
- Seq:06