Grounding cables, b.safe
Grounding cable with clip and ring cable lug M6, 1,5 m
Codice catalogo 590-0338
Nome del fornitore: Bohlender
Vai a
Product Details & Documents
For earth connection of vessels and components made of static dissipative materials.
- Choose the suitable connection to earth that we offer in our range of grounding cables with different connectors
- Customised versions can be offered on request
- Suitable grounding accessories for b.safe products
The scope of delivery of the ring cable lug includes screw, nut and washer.
Specifiche
- Descrizione:Grounding cable with clip and ring cable lug M6
- Lunghezza:1,5 m
- Seq:04