Orders LinkContactUsLinkComponent
Capillary cutter, b.safe
Capillary cutter, b.safe
Replacement blades for capillary cutter for tubing up to Ext.Ø 8 mm
Codice catalogo 590-0336
Nome del fornitore:   Bohlender
Capillary cutter, b.safe
Codice catalogo 590-0336
Nome del fornitore:  Bohlender
Capillary cutter, b.safe
Capillary cutter, b.safe
Replacement blades for capillary cutter for tubing up to Ext.Ø 8 mm
Price
13.70 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Ideal for cutting capillaries and elastic tubes without reinforcement.
  • Capillary cutter for up to tubing Ext.Ø 8 mm with guide bores
  • High qualitiy capillary cutter
  • Replacement blades available
The guide bores help to get a clean cut of the capillaries.
Specifiche
  • Descrizione:
    Replacement blades for capillary cutter for tubing up to Ext.Ø 8 mm
  • Seq:
    03