Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
3',5'-Dihydroxyacetophenone 98%

3',5'-Dihydroxyacetophenone 98%

Bottle, 100 g
Catalog # APOSOR952849-100G
Supplier:   Apollo Scientific
3',5'-Dihydroxyacetophenone 98%
Catalog # APOSOR952849-100G
Supplier:  Apollo Scientific
CAS Number:  51863-60-6
3',5'-Dihydroxyacetophenone 98%

Bottle, 100 g

CAS Number:  51863-60-6
Price
SFr. 108.00
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1-(3,5-Dihydroxyphenyl)ethan-1-one, 1-(3,5-Dihydroxyphenyl)ethanone
  • CAS Number:51863-60-6
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00002290
  • Molecular Weight:152.15 g/mol
  • Storage Temperature:Ambiente
Specifications

Specifications

  • Tipo di conf.:
    Bottle
  • Conf:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.