Orders LinkContactUsLinkComponent
D(-)-Isoleucine
D(-)-Isoleucine
Bottle, 5 g
Catalog # APOSOR36341-5G
Supplier:   Apollo Scientific
D(-)-Isoleucine
Catalog # APOSOR36341-5G
Supplier:  Apollo Scientific
CAS Number:  319-78-8
D(-)-Isoleucine
Bottle, 5 g
CAS Number:  319-78-8
Price
SFr. 856.00
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  (2R,3R)-(-)-Isoleucine, H-D-Ile-OH
  • CAS Number:319-78-8
  • Formula:H₃CCH₂CH(CH₃)CH(NH₂)COOH
  • MDL Number:MFCD00064221
  • Molecular Weight:131.17 g/mol
  • Melting Pt:265…274 °C
  •  :Density: 1.224 g/cm³ (20 °C)
  • Storage Temperature:Ambiente
Specifications
Specifications
  • Tipo di conf.:
    Bottle
  • Conf:
    5 g