Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Fumaroyl chloride 95%

Fumaroyl chloride 95%

Bottle, 25 g
Catalog # APOSOR923090-25G
Supplier:   Apollo Scientific
Fumaroyl chloride 95%
Catalog # APOSOR923090-25G
Supplier:  Apollo Scientific
CAS Number:  627-63-4
Fumaroyl chloride 95%

Bottle, 25 g

CAS Number:  627-63-4
Price
SFr. 156.00
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Fumaryl chloride
  • CAS Number:627-63-4
  • Formula:C₄H₂Cl₂O₂
  • MDL Number:MFCD00000733
  • Molecular Weight:152.96 g/mol
  • Storage Temperature:Ambiente
Specifications

Specifications

  • Tipo di conf.:
    Bottle
  • Conf:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.