Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
3-Nitro-L-tyrosine 98%

3-Nitro-L-tyrosine 98%

Bottle, 25 g
Catalog # APOSOR9954-25G
Supplier:   Apollo Scientific
3-Nitro-L-tyrosine 98%
Catalog # APOSOR9954-25G
Supplier:  Apollo Scientific
CAS Number:  621-44-3
3-Nitro-L-tyrosine 98%

Bottle, 25 g

CAS Number:  621-44-3
Price
SFr. 163.00
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  H-Tyr(3-NO2)-OH, 3-Nitro-L-tyrosine
  • CAS Number:621-44-3
  • Formula:C₉H₁₀N₂O₅
  • MDL Number:MFCD00007123
  • Molecular Weight:226.3 g/mol
  • Storage Temperature:Ambiente
Specifications

Specifications

  • Tipo di conf.:
    Bottle
  • Conf:
    25 g

Specifications