Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
3-Chlorophenethyl bromide 97%

3-Chlorophenethyl bromide 97%

Bottle, 25 g
Catalog # APOSOR952414-25G
Supplier:   Apollo Scientific
3-Chlorophenethyl bromide 97%
Catalog # APOSOR952414-25G
Supplier:  Apollo Scientific
CAS Number:  16799-05-6
3-Chlorophenethyl bromide 97%

Bottle, 25 g

CAS Number:  16799-05-6
Price
SFr. 584.00
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  3-Chlorophenethyl Bromide
  • CAS Number:16799-05-6
  • Formula:C₈H₈BrCl
  • MDL Number:MFCD04117465
  • Molecular Weight:219.51 g/mol
  • Storage Temperature:Ambiente
Specifications

Specifications

  • Tipo di conf.:
    Bottle
  • Conf:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.