Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
2'-Bromoacetophenone 98%

2'-Bromoacetophenone 98%

Bottle, 500 g
Catalog # APOSOR2031-500G
Irritant
Supplier:   Apollo Scientific
2'-Bromoacetophenone 98%
Catalog # APOSOR2031-500G
Supplier:  Apollo Scientific
CAS Number:  2142-69-0
2'-Bromoacetophenone 98%

Bottle, 500 g

CAS Number:  2142-69-0
Price
607.00 SFr.
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  o-Bromoacetophenone
  • CAS Number:2142-69-0
  • Formula:C₈H₇BrO
  • MDL Number:MFCD00000067
  • Molecular Weight:199.05 g/mol
  • Density:1,476 g/cm³ (25 °C)
  • Flash Pt:>110 °C
  • Storage Temperature:Ambiente
  • EINECS:218-398-1
Specifications

Specifications

  • Tipo di conf.:
    Bottle
  • Conf:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.