ContactUsLinkComponent
Human CD16b NA2, His Tag SPR  BLI verified CDB-H5222 is expressed from human 293 cells HEK293. It contains AA Gly 17 - Ser 200 Acce ssion O75015-1. null
Codice catalogo ACRBCDB-H5222-50UG
Nome del fornitore:  ACRO Biosystems
Human CD16b NA2, His Tag SPR  BLI verified CDB-H5222 is expressed from human 293 cells HEK293. It contains AA Gly 17 - Ser 200 Acce ssion O75015-1. null
Codice catalogo ACRBCDB-H5222-50UG
Nome del fornitore:  ACRO Biosystems
Frequently Bought Together