Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 SFr.
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Human PD-L1, His Tag PD1-H5229 is expressed from human 293 cells HEK293. It contains AA Phe 19 - Arg 238 Accession NP_054862.1 . 1 * 100 µG
Catalog # ACRBPD1-H5229100UG
Supplier:
ACRO Biosystems
Expand image
Human PD-L1, His Tag PD1-H5229 is expressed from human 293 cells HEK293. It contains AA Phe 19 - Arg 238 Accession NP_054862.1 . 1 * 100 µG
Catalog # ACRBPD1-H5229100UG
Supplier:
ACRO Biosystems
Frequently Bought Together
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
VWR®, Provette e strip per PCR, 0,2 ml
Supplier: VWR Collection
VWR®, Pipette sierologiche, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Provette per centrifuga, Falcon®
Supplier: Corning
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR®, Cucchiai per campionamento
Supplier: VWR Collection
VWR®, Pipette Pasteur, Standard Line
Supplier: VWR Collection
VWR®, Divaricatori, a L, monouso
Supplier: VWR Collection
VWR®, pipette di trasferimento, alte prestazioni
Supplier: VWR Collection
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
Provette per microcentrifuga Eppendorf® Protein LoBind®
Supplier: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF