ContactUsLinkComponent
Biotinylated Human BCMA, His,Avitag BCA-H82E4 is expressed from human 293 cells HEK293. It contains AA Met 1 - Ala 54 Accession Q02 223-1. 1 * 25 µG
Catalog # ACRBBCA-H82E4-25UG
Supplier:  ACRO Biosystems
Biotinylated Human BCMA, His,Avitag BCA-H82E4 is expressed from human 293 cells HEK293. It contains AA Met 1 - Ala 54 Accession Q02 223-1. 1 * 25 µG
Catalog # ACRBBCA-H82E4-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together