Rhesus macaque CD32a, His Tag SPR BLI verified CDA-R52H4 is expressed from human 293 cells HEK293. It contains AA Gln 34 - Ile 217 Accession NP_001244229.1. 1 * 50 µG
Rhesus macaque CD32a, His Tag SPR BLI verified CDA-R52H4 is expressed from human 293 cells HEK293. It contains AA Gln 34 - Ile 217 Accession NP_001244229.1. 1 * 50 µG