Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Human recombinant Her2 (from HEK293 cells)

100 µG
  ACRBHE2-H52H4100UG
Marketsource
Human recombinant Her2 (from HEK293 cells)
  ACRBHE2-H52H4100UG
Marketsource
 :  
Human recombinant Her2 (from HEK293 cells)

100 µG

 :  
 
SFr. 360.00
   
1 * 100 µG
 
 

 

  • Conf:
    100 µG
  • Protein/peptide name:
    Her2
  • Purezza:
    >90% as determined by SDS-PAGE.
  • Protein synonyms:
    NEU|MLN19|TKR1|ERBB2|CD340|HER2|HER-2/neu|NGL
  • Specie:
    Human
  • Coniugazione:
    Non coniugato
  • Sorgente luminosa:
    HEK293 cells
  • Peso molecolare:
    18,5 kDa
  • Cat. No.:
    ACRBHE2-H52H4100UG
  • Condizioni di conservazione:
    –20 °C
  • Contenuto di endotossine:
    Less than 1,0 EU per μg by the LAL method.
  • Protein/peptide type:
    Recombinant
  • Livello di endotossine:
    Low
  • ID gene:
    P04626-1
  • Formulazione:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Temperatura di spedizione:
    RT
  • Sequenza tag:
    His Tag