FITC-Labeled Human GUCY2C, His Tag GUC-HF2H8 is expressed from human 293 cells HEK293. It contains AA Ser 24 - Gln 430 Accession P2 5092-1. It is the FITC labeled form of Human GUCY2C, His Tag GUC-H52H5. 1 * 25 µG
FITC-Labeled Human GUCY2C, His Tag GUC-HF2H8 is expressed from human 293 cells HEK293. It contains AA Ser 24 - Gln 430 Accession P2 5092-1. It is the FITC labeled form of Human GUCY2C, His Tag GUC-H52H5. 1 * 25 µG