Cynomolgus GUCY2C, His Tag GUC-C52H6 is expressed from human 293 cells HEK293. It contains AA Ser 24 - Gln 430 Accession XP_0055702 70.1. 1 * 100 µG
Codice catalogo ACRBGUC-C52H6100UG
Nome del fornitore: ACRO Biosystems
Frequently Bought Together