ContactUsLinkComponent
SARS-CoV-2 S1 protein CTD, His Tag MALS-verified S1D-C52H3 is expressed from human 293 cells HEK293. It contains AA Asn 334 - Pro 52 7 Accession QHD43416.1. 1 * 100 µG
  ACRBS1D-C52H3100UG
SARS-CoV-2 S1 protein CTD, His Tag MALS-verified S1D-C52H3 is expressed from human 293 cells HEK293. It contains AA Asn 334 - Pro 52 7 Accession QHD43416.1. 1 * 100 µG
  ACRBS1D-C52H3100UG
Frequently Bought Together