ContactUsLinkComponent
SARS-CoV-2 S protein, His Tag, Super stable trimer SPN-C52H9 is the ectodomain of SARS-CoV-2 S protein which contains AA Val 16 - Pr o 1213 Accession  QHD43416.1. The recombinant protein is expressed from human 293 cells HEK293 with T4 fibritin trime 1 * 50 µG
  ACRBSPN-C52H9-50UG
SARS-CoV-2 S protein, His Tag, Super stable trimer SPN-C52H9 is the ectodomain of SARS-CoV-2 S protein which contains AA Val 16 - Pr o 1213 Accession  QHD43416.1. The recombinant protein is expressed from human 293 cells HEK293 with T4 fibritin trime 1 * 50 µG
  ACRBSPN-C52H9-50UG
Frequently Bought Together