ContactUsLinkComponent
SARS-CoV-2 S protein D614G, His Tag, Super stable trimer SPN-C52H3 is the ectodomain of SARS-CoV-2 S protein that contains AA Val 16 - Pro 1213 Accession QHD43416.1 and D614G mutation, which has become increasingly common in SARS-CoV-2 viruses from l 1 * 50 µG
Codice catalogo ACRBSPN-C52H3-50UG
Nome del fornitore:  ACROBIOSYSTEMS
SARS-CoV-2 S protein D614G, His Tag, Super stable trimer SPN-C52H3 is the ectodomain of SARS-CoV-2 S protein that contains AA Val 16 - Pro 1213 Accession QHD43416.1 and D614G mutation, which has become increasingly common in SARS-CoV-2 viruses from l 1 * 50 µG
Codice catalogo ACRBSPN-C52H3-50UG
Nome del fornitore:  ACROBIOSYSTEMS
Frequently Bought Together