ContactUsLinkComponent
FITC-Labeled Human Syndecan-1, His Tag SD1-HF228 is expressed from human 293 cells HEK293. It contains AA Gln 23 - Gly 254 Accessio n P18827-1. 1 * 25 µG
Codice catalogo ACRBSD1-HF228-25UG
Nome del fornitore:  ACROBIOSYSTEMS
FITC-Labeled Human Syndecan-1, His Tag SD1-HF228 is expressed from human 293 cells HEK293. It contains AA Gln 23 - Gly 254 Accessio n P18827-1. 1 * 25 µG
Codice catalogo ACRBSD1-HF228-25UG
Nome del fornitore:  ACROBIOSYSTEMS
Frequently Bought Together