ContactUsLinkComponent
FITC-Labeled Human IL-3 R alpha, His Tag ILA-HF2H9 is expressed from human 293 cells HEK293. It contains AA Thr 19 - Arg 305  Access ion P26951-1. 1 * 200 µG
Catalog # ACRBILA-HF2H9200UG
Supplier:  ACROBIOSYSTEMS
FITC-Labeled Human IL-3 R alpha, His Tag ILA-HF2H9 is expressed from human 293 cells HEK293. It contains AA Thr 19 - Arg 305  Access ion P26951-1. 1 * 200 µG
Catalog # ACRBILA-HF2H9200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together