FITC-Labeled Human CD5, His Tag CD5-H52H2 is expressed from human 293 cells HEK293. It contains AA Arg 25 - Pro 372 Accession P0612 7-1. It is the FITC labeled form of Human CD5, His Tag CD5-H52H5. 1 * 25 µG
Codice catalogo ACRBCD5-H52H2-25UG
Nome del fornitore: ACROBIOSYSTEMS
Frequently Bought Together