ContactUsLinkComponent
Cynomolgus CD3ECD3G Heterodimer Protein, Fc TagFc Tag CDG-C5257 is expressed from human 293 cells HEK293. It contains AA Gln 22 - As p 117 CD3E Gln 23 - Thr 113 CD3G Accession Q95LI5-1 CD3E Q95LI7-1 CD3G. 1 * 100 µG
Codice catalogo ACRBCDG-C5257100UG
Nome del fornitore:  ACROBIOSYSTEMS
Cynomolgus CD3ECD3G Heterodimer Protein, Fc TagFc Tag CDG-C5257 is expressed from human 293 cells HEK293. It contains AA Gln 22 - As p 117 CD3E Gln 23 - Thr 113 CD3G Accession Q95LI5-1 CD3E Q95LI7-1 CD3G. 1 * 100 µG
Codice catalogo ACRBCDG-C5257100UG
Nome del fornitore:  ACROBIOSYSTEMS
Frequently Bought Together