Human IL17RA IL17RC Protein, Fc TagFc Tag ILC-H5257 is expressed from human 293 cells HEK293. It contains AA Leu 33 - Trp 320 IL-17 RA Leu 21 - His 465 IL-17RC Accession Q96F46-1 IL-17RA NP_703190.1 IL-17RC. 1 * 1 mg
Catalog # ACRBILC-H5257-1MG
Supplier: ACROBIOSYSTEMS
Frequently Bought Together