ContactUsLinkComponent
FITC-Labeled Human EpCAM, Fc Tag EPM-HF255 is expressed from human 293 cells HEK293. It contains AA Gln 24 - Lys 265 Accession AAH1 4785.1. 1 * 25 µG
Catalog # ACRBEPM-HF255-25UG
Supplier:  ACRO Biosystems
FITC-Labeled Human EpCAM, Fc Tag EPM-HF255 is expressed from human 293 cells HEK293. It contains AA Gln 24 - Lys 265 Accession AAH1 4785.1. 1 * 25 µG
Catalog # ACRBEPM-HF255-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together