ContactUsLinkComponent
FITC-Labeled Human EpCAM, Fc Tag EPM-HF255 is expressed from human 293 cells HEK293. It contains AA Gln 24 - Lys 265 Accession AAH1 4785.1. 1 * 200 µG
Codice catalogo ACRBEPM-HF255200UG
Nome del fornitore:  ACROBIOSYSTEMS
FITC-Labeled Human EpCAM, Fc Tag EPM-HF255 is expressed from human 293 cells HEK293. It contains AA Gln 24 - Lys 265 Accession AAH1 4785.1. 1 * 200 µG
Codice catalogo ACRBEPM-HF255200UG
Nome del fornitore:  ACROBIOSYSTEMS
Frequently Bought Together