ContactUsLinkComponent
FITC-Labeled Human EpCAM, His Tag EPM-HF225 is expressed from human 293 cells HEK293. It contains AA Gln 24 - Lys 265 Accession AAH 14785.1. 1 * 200 µG
Codice catalogo ACRBEPM-HF225200UG
Nome del fornitore:  ACROBIOSYSTEMS
FITC-Labeled Human EpCAM, His Tag EPM-HF225 is expressed from human 293 cells HEK293. It contains AA Gln 24 - Lys 265 Accession AAH 14785.1. 1 * 200 µG
Codice catalogo ACRBEPM-HF225200UG
Nome del fornitore:  ACROBIOSYSTEMS
Frequently Bought Together