Anti-SARS-CoV-2 RBD Potent Neutralizing Antibody, Chimeric mAb, Human IgG1 AM128 SPD-M128 is a chimeric monoclonal antibody combinin g the constant domains of the human IgG1 molecule with mouse variable regions. Pseudovirus assay shows that t 1 * 100 µG
ACRBSPD-M128-100UG
Frequently Bought Together