Auftragserfassung
Switzerland
ContactUsLinkComponent
FITC-Labeled Human CEACAM-5, His Tag CE5-HF254 is expressed from human 293 cells HEK293. It contains AA Lys 35 - Ala 685 Accession P06731-1. It is the FITC labeled form of Human CEACAM-5, His Tag Cat. No.CE5-H5226. 1 * 200 µG
  ACRBCE5-HF254200UG
FITC-Labeled Human CEACAM-5, His Tag CE5-HF254 is expressed from human 293 cells HEK293. It contains AA Lys 35 - Ala 685 Accession P06731-1. It is the FITC labeled form of Human CEACAM-5, His Tag Cat. No.CE5-H5226. 1 * 200 µG
  ACRBCE5-HF254200UG
Frequently Bought Together