ContactUsLinkComponent
FITC-Labeled Human NKG2D, Fc Tag NKD-HF224 is expressed from human 293 cells HEK293. It contains AA Ile 73 - Val 216 Accession P267 18-1. It is the biotinylated form of Human NKG2D, Fc Tag HPLC-verified Cat. No.NKD-H5265. 1 * 200 µG
Codice catalogo ACRBNKD-HF224200UG
Nome del fornitore:  ACROBIOSYSTEMS
FITC-Labeled Human NKG2D, Fc Tag NKD-HF224 is expressed from human 293 cells HEK293. It contains AA Ile 73 - Val 216 Accession P267 18-1. It is the biotinylated form of Human NKG2D, Fc Tag HPLC-verified Cat. No.NKD-H5265. 1 * 200 µG
Codice catalogo ACRBNKD-HF224200UG
Nome del fornitore:  ACROBIOSYSTEMS
Frequently Bought Together