Auftragserfassung
Switzerland
ContactUsLinkComponent
Cynomolgus Rhesus macaque CD19 20-292, His Tag CD9-C5221 is expressed from human 293 cells HEK293. It contains AA Pro 20 - Lys 292 Accession F7F486-1. 1 * 100 µG
Catalog # ACRBCD9-C5221100UG
Supplier:  ACRO Biosystems
Cynomolgus Rhesus macaque CD19 20-292, His Tag CD9-C5221 is expressed from human 293 cells HEK293. It contains AA Pro 20 - Lys 292 Accession F7F486-1. 1 * 100 µG
Catalog # ACRBCD9-C5221100UG
Supplier:  ACRO Biosystems
Frequently Bought Together