ContactUsLinkComponent
Human CD133 Full Length, His Tag Nanodisc CD3-H52H1 is expressed from human 293 cells HEK293. It contains AA Gly 20 - His 865 Access ion O43490-1. 1 * 100 µG
Catalog # ACRBCD3-H52H1100UG
Supplier:  ACRO Biosystems
Human CD133 Full Length, His Tag Nanodisc CD3-H52H1 is expressed from human 293 cells HEK293. It contains AA Gly 20 - His 865 Access ion O43490-1. 1 * 100 µG
Catalog # ACRBCD3-H52H1100UG
Supplier:  ACRO Biosystems
Frequently Bought Together