ContactUsLinkComponent
Human TREM2, Mouse IgG2a Fc Tag TR2-H5256 is expressed from human 293 cells HEK293. It contains AA His 19 - Ser 174 Accession Q9NZC 2-1. 1 * 1 mg
Codice catalogo ACRBTR2-H5256-1MG
Nome del fornitore:  ACRO Biosystems
Human TREM2, Mouse IgG2a Fc Tag TR2-H5256 is expressed from human 293 cells HEK293. It contains AA His 19 - Ser 174 Accession Q9NZC 2-1. 1 * 1 mg
Codice catalogo ACRBTR2-H5256-1MG
Nome del fornitore:  ACRO Biosystems
Frequently Bought Together