ContactUsLinkComponent
Human CD3 gamma, Fc Tag CDA-H5254 is expressed from human 293 cells HEK293. It contains AA Gln 23 - Ser 116 Accession P09693-1. 1 * 1 mg
Codice catalogo ACRBCDA-H5254-1MG
Nome del fornitore:  ACRO Biosystems
Human CD3 gamma, Fc Tag CDA-H5254 is expressed from human 293 cells HEK293. It contains AA Gln 23 - Ser 116 Accession P09693-1. 1 * 1 mg
Codice catalogo ACRBCDA-H5254-1MG
Nome del fornitore:  ACRO Biosystems
Frequently Bought Together