ContactUsLinkComponent
FITC-Labeled Human CD47, His Tag CD7-HF2H3 is expressed from human 293 cells HEK293. It contains AA Gln 19 - Pro 139 Accession NP_9 42088. 1 * 25 µG
Codice catalogo ACRBCD7-HF2H3-25UG
Nome del fornitore:  ACROBIOSYSTEMS
FITC-Labeled Human CD47, His Tag CD7-HF2H3 is expressed from human 293 cells HEK293. It contains AA Gln 19 - Pro 139 Accession NP_9 42088. 1 * 25 µG
Codice catalogo ACRBCD7-HF2H3-25UG
Nome del fornitore:  ACROBIOSYSTEMS
Frequently Bought Together