FITC-Labeled Human CD47, His Tag CD7-HF2H3 is expressed from human 293 cells HEK293. It contains AA Gln 19 - Pro 139 Accession NP_9 42088. 1 * 200 µG
Codice catalogo ACRBCD7-HF2H3200UG
Nome del fornitore: ACROBIOSYSTEMS
Frequently Bought Together