SARS-CoV-2 S protein RBD K417N, E484K, N501Y, His Tag SPD-C52Hp is expressed from human 293 cells HEK293. It contains AA Arg 319 - Lys 537 Accession QHD43416.1 K417N, E484K, N501Y. K417N E484K N501Y mutations were identified in the SARS-CoV-2 var 20 1 * 100 µG
Codice catalogo ACRBSPD-C52HP100UG
Nome del fornitore: ACROBIOSYSTEMS
Frequently Bought Together