ContactUsLinkComponent
SARS-CoV-2 S protein RBD K417N, His Tag SPD-C52Hs is expressed from human 293 cells HEK293. It contains AA Arg 319 - Lys 537 Access ion QHD43416.1 K417N. 1 * 100 µG
Codice catalogo ACRBSPD-C52HS100UG
Nome del fornitore:  ACRO Biosystems
SARS-CoV-2 S protein RBD K417N, His Tag SPD-C52Hs is expressed from human 293 cells HEK293. It contains AA Arg 319 - Lys 537 Access ion QHD43416.1 K417N. 1 * 100 µG
Codice catalogo ACRBSPD-C52HS100UG
Nome del fornitore:  ACRO Biosystems
Frequently Bought Together