SARS-CoV-2 S2 protein A701V, His Tag S2N-C52Hc is expressed from human 293 cells HEK293. It contains AA Ser 686 - Pro 1213 Accession QHD43416.1 A701V, F817P, A892P, A899P, A942P, K986P, V987P). The recombinant protein is expressed from human 293 ce 1 * 1 mg
Catalog # ACRBS2N-C52HC-1MG
Supplier: ACRO Biosystems
Frequently Bought Together