Orders LinkContactUsLinkComponent
Industrial visor range, EVOGuard®
Industrial visor range, EVOGuard®
Clear visor, 185 mm, anti-mist, anti-scratch
Codice catalogo JSPAADA250-060-000
Nome del fornitore:   JSP
Industrial visor range, EVOGuard®
Codice catalogo JSPAADA250-060-000
Nome del fornitore:  JSP
Industrial visor range, EVOGuard®
Industrial visor range, EVOGuard®
Clear visor, 185 mm, anti-mist, anti-scratch
Price
SFr. 42.20
Log in per disponibilità
1 * 1 ST
Product Details & Documents
A range of high performance industrial visors, fully compatible with EVOGuard® browguard systems and EVO® helmet models.
  • Polycarbonate clear visor
  • Optical class 1 visor
  • Visors filter harmful UV radiation (UV 2C-1.2 protection)
  • Offering up to A-rated impact resistance
The optical class 1 visor offers a panoramic view with minimal distortion, ideal for comfortable all day wear. EVOGuard® visors maximise compatibility with JSP half masks, disposable respirators, prescription glasses, and the Sonis® helmet-mounted ear defender range. Visor filters harmful UV radiation to protect the eyes whilst also offering high light transmission and enhanced colour recognition.
Five high performance visors offering up to A rated impact resistance when fitted to a safety helmet and fully compatible with EVOGuard® browguard for medium energy impact protection. EVOGuard® visors provide comprehensive facial coverage without limiting movement.
Certifications: EN 170; EN 166.
Specifiche
  • Descrizione:
    Clear visor, 185 mm, anti-mist, anti-scratch
  • Seq:
    02