ContactUsLinkComponent
Pt100 4-wire temperature probe, Temperature range -50 °C.. +105 °C, DIN A, plastic material, cable length 5m, with M12 connector 1 * 1 ST
Catalog # ELPR802243
Supplier:  ELPRO-BUCHS
Pt100 4-wire temperature probe, Temperature range -50 °C.. +105 °C, DIN A, plastic material, cable length 5m, with M12 connector 1 * 1 ST
Catalog # ELPR802243
Supplier:  ELPRO-BUCHS
Frequently Bought Together