[EN]TIP EJECTOR 20 - 50 -100 CLICK-2-FI 1 * 1 pezzi
VWRISP19052
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
VWR®, Accessori per pipette meccaniche
: VWR Collection
Acqua, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
: VWR Chemicals
VWR®, Mini vortex mixer
: VWR Collection
Acqua, HiPerSolv CHROMANORM® per HPLC
: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
: VWR Chemicals
Etanolo, GPR RECTAPUR®
: VWR Chemicals
Accessori per accu-jet® pro
: Brand
VWR®, Pipette sierologiche, Standard Line
: VWR Collection