Adapter kit, Neuros™
Neuros™ Adapter kit for select 700/1700 syringes, 3, 33
Codice catalogo HAMI65460-04
Nome del fornitore: HAMILTON BONADUZ
Vai a
Product Details & Documents
The Neuros™ adapter kit attaches to any 5,0 to 100 µl removable needle (RN) syringe.
- Gas sterilisable, not autoclavable
- Stainless steel, borosilicate glass fluid path
The needle sleeves come in two versions, one with a blind stop and one without. The sleeve with a stop is designed for applications where it is desirable to limit the penetration depth of the needle, as is the case with a cannulated animal. The sleeve without a stop supports the needle but does not obstruct the view of the injection target which is ideal for microsurgeries using a stereotaxic holder.
Delivery information: The kit includes a glass adapter, two needle sleeves and a six pack of 33 gauge needles.
Specifiche
- Gauge:33
- Tipo punta:3
- Descrizione:Neuros™ Adapter kit for select 700/1700 syringes
- Seq:02