Orders LinkContactUsLinkComponent
Adapter kit, Neuros™
Adapter kit, Neuros™
Neuros™ Adapter kit for select 700/1700 syringes, 3, 33
Codice catalogo HAMI65460-04
Nome del fornitore:   HAMILTON BONADUZ
Adapter kit, Neuros™
Codice catalogo HAMI65460-04
Nome del fornitore:  HAMILTON BONADUZ
Adapter kit, Neuros™
Neuros™ Adapter kit for select 700/1700 syringes, 3, 33
Price
SFr. 404.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
The Neuros™ adapter kit attaches to any 5,0 to 100 µl removable needle (RN) syringe.
  • Gas sterilisable, not autoclavable
  • Stainless steel, borosilicate glass fluid path
The needle sleeves come in two versions, one with a blind stop and one without. The sleeve with a stop is designed for applications where it is desirable to limit the penetration depth of the needle, as is the case with a cannulated animal. The sleeve without a stop supports the needle but does not obstruct the view of the injection target which is ideal for microsurgeries using a stereotaxic holder.
Delivery information: The kit includes a glass adapter, two needle sleeves and a six pack of 33 gauge needles.
Specifiche
  • Gauge:
    33
  • Tipo punta:
    3
  • Descrizione:
    Neuros™ Adapter kit for select 700/1700 syringes
  • Seq:
    02