Pfu DNA polymerase
Cloned Pfu DNA polymerase, Cloned Pfu DNA polymerase (2.5 U/µl), 10X cloned Pfu buffer, 1000 U, 1
Codice catalogo AGLS600159
Nome del fornitore: AGILENT
Vai a
Product Details & Documents
High-quality polymerases for difficult to amplify DNA targets.
- Proofreading DNA polymerase isolated from Pyrococcus furiosus
- Ideal choice for many techniques requiring high-fidelity DNA synthesis by PCR
- Blunt ends
Pfu DNA Polymerases includes Pfu DNA Polymerase, a proofreading DNA polymerase isolated from Pyrococcus furiosus.
The portfolio also features the recombinant Exo- Pfu DNA Polymerase (a genetically engineered mutant of the cloned Pfu DNA format) and the PfuTurbo DNA Polymerase with an optional alternative detergent.
Caution: For research use only. Not for use in diagnostic procedures.
Specifiche
Specifiche
- Conf:1
- Descrizione:Cloned Pfu DNA polymerase
- Taglia:1000 U
- Details:Cloned Pfu DNA polymerase (2.5 U/µl), 10X cloned Pfu buffer
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



