Orders LinkContactUsLinkComponent
Pfu DNA polymerase
Native Pfu DNA polymerase (2.5 U/µl), 10X native Pfu buffer
  AGLS600135
 :   AGILENT
Pfu DNA polymerase
  AGLS600135
 :  AGILENT
 :  
Pfu DNA polymerase
Native Pfu DNA polymerase (2.5 U/µl), 10X native Pfu buffer
 :  
1 * 1 ST
High-quality polymerases for difficult to amplify DNA targets.
  • Proofreading DNA polymerase isolated from Pyrococcus furiosus
  • Ideal choice for many techniques requiring high-fidelity DNA synthesis by PCR
  • Blunt ends
Pfu DNA Polymerases includes Pfu DNA Polymerase, a proofreading DNA polymerase isolated from Pyrococcus furiosus.
The portfolio also features the recombinant Exo- Pfu DNA Polymerase (a genetically engineered mutant of the cloned Pfu DNA format) and the PfuTurbo DNA Polymerase with an optional alternative detergent.
 : For research use only. Not for use in diagnostic procedures.
  • Details:
    Native Pfu DNA polymerase (2.5 U/µl), 10X native Pfu buffer