AccuScript high-fidelity reverse transcriptase
AccuScript reverse transcriptase, Engineered to deliver the highest transcription accuracy while promoting full-length cDNA synthesis, yield, and RT-PCR performance, 50 reactions, 1
Codice catalogo AGLS600089
Nome del fornitore: AGILENT
AccuScript high-fidelity reverse transcriptase
Codice catalogo AGLS600089
Nome del fornitore: AGILENT
Numero CAS:
Vai a
Product Details & Documents
AccuScript High-Fidelity Reverse Transcriptase delivers the highest RT accuracy currently available.
- MMLV-RT derivative combined with 3 to 5’ exonuclease reduces errors
- High yields of full length cDNA up to 20 kb;RT-PCR kit synthesises cDNA from 10 to 1000 ng of total RNA or 0,1 to 10 ng of poly(A)+ RNA
- Stringent quality control procedures ensure high yields of premium cDNA
AccuScript High-Fidelity Reverse Transcriptase (RT) delivers 3,7 times more accuracy than other commercially available RTs and 6,6 times more accuracy than AMV RT. The AccuScript PfuUltra II RT-PCR kit provides a complete system in applications that demand high fidelity, specificity, and throughput.
Attenzione: For research use only. Not for use in diagnostic procedures.
Specifiche
Specifiche
- Conf:1
- Descrizione:AccuScript reverse transcriptase
- Details:Engineered to deliver the highest transcription accuracy while promoting full-length cDNA synthesis, yield, and RT-PCR performance
- No. of reactions:50 reactions