Thermoblock, SmartBlocks™
SmartBlock™ cryo thaw, thermoblock for 24 cryo tubes, 1,5 – 2,0 ml, all base shapes
Codice catalogo 460-0366
Nome del fornitore: EPPENDORF
Thermoblock, SmartBlocks™
Codice catalogo 460-0366
Nome del fornitore: EPPENDORF
Vai a
Product Details & Documents
Exchangeable in just seconds. Individually sensor-controlled SmartBlocks are your solution for maintaining accurate, uniform temperatures. A wide variety of these flexible thermoblocks for all common laboratory vessels support a broad range of applications - for use with Eppendorf ThermoMixer® C and ThermoStat C.
- Optimised block design supports optimal temperature transfer to sample to achieve excellent results
- Eppendorf SmartBlock-specific calibration by integrated sensor technology maximizes temperature accuracy and homogeneity
- Insulated thermoblocks for ergonomic operation - you won't burn your fingers (PhysioCare Concept®)
- Automatic block recognition for safe handling
- Optimised block design ensures maximum temperature transfer to the sample
- Eppendorf QuickRelease technology enables fast and simple block exchange – no need for tools
Every Eppendorf SmartBlock is individually inspected in production for temperature accuracy and includes a serial-number specific certificate for documentation purpose.
Documents
EPPE5362000.035_TDS_EN
Scheda dati
Specifiche
- Descrizione:SmartBlock™ cryo thaw, thermoblock for 24 cryo tubes, 1,5 – 2,0 ml, all base shapes
- Seq:15
- Cat. No.:460-0366
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:Renewable Energy Certificate, ISO 14001